ATC311766536 Atlantic Research Chemicals Ltd

2-Bromofuran

  • MDL number MFCD00216609
  • Cas Number : 584-12-3
  • Molecular Weight : 146.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA00401
Availability details for
Quantity
  • Availability
  • Add to Cart