ATC311766544 Atlantic Research Chemicals Ltd

2-Bromo-5-methyl-1,3-thiazole

  • MDL number MFCD08460610
  • Cas Number : 41731-23-1
  • Molecular Weight : 178.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA00602
Availability details for
Quantity
  • Availability
  • Add to Cart