ATC311766547 Atlantic Research Chemicals Ltd

2-Amino-5-bromo-1,3-thiazole

  • MDL number MFCD00016893
  • Cas Number : 3034-22-8
  • Molecular Weight : 179.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA00605
Availability details for
Quantity
  • Availability
  • Add to Cart