ATC311766600 Atlantic Research Chemicals Ltd

5-Bromo-1H-imidazole

  • MDL number MFCD02728863
  • Cas Number : 2302-25-2
  • Molecular Weight : 146.975

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=HA01015
Availability details for
Quantity
  • Availability
  • Add to Cart