ATC311766612 Atlantic Research Chemicals Ltd

3-Bromo-5-methylpyrazin-2-amine

  • MDL number MFCD09750037
  • Cas Number : 74290-65-6
  • Molecular Weight : 188.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA01112
Availability details for
Quantity
  • Availability
  • Add to Cart