ATC311766637 Atlantic Research Chemicals Ltd

2-Chloro-4-piperidin-1-ylpyrimidine

  • MDL number MFCD01912571
  • Cas Number : 5429-00-5
  • Molecular Weight : 197.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA01222
Availability details for
Quantity
  • Availability
  • Add to Cart