ATC311766640 Atlantic Research Chemicals Ltd

2,4-Dibromopyrimidine

  • MDL number MFCD06798232
  • Cas Number : 3921-01-5
  • Molecular Weight : 237.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA01225
Availability details for
Quantity
  • Availability
  • Add to Cart