ATC311766760 Atlantic Research Chemicals Ltd

4-Bromo-3-(trifluoromethyl)quinoline

  • MDL number MFCD08458128
  • Cas Number : 590371-97-4
  • Molecular Weight : 276.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA02605
Availability details for
Quantity
  • Availability
  • Add to Cart