ATC311766763 Atlantic Research Chemicals Ltd

2-Chloro-3-methylquinoline

  • MDL number MFCD00100265
  • Cas Number : 57876-69-4
  • Molecular Weight : 177.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA02608
Availability details for
Quantity
  • Availability
  • Add to Cart