ATC311766782 Atlantic Research Chemicals Ltd

5-Bromoquinoxalin-6-amine

  • MDL number MFCD00799024
  • Cas Number : 50358-63-9
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA02803
Availability details for
Quantity
  • Availability
  • Add to Cart