ATC311766823 Atlantic Research Chemicals Ltd

3-Chloro-1,2-benzisoxazole

  • MDL number MFCD03407314
  • Cas Number : 16263-52-8
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA05201
Availability details for
Quantity
  • Availability
  • Add to Cart