ATC311766854 Atlantic Research Chemicals Ltd

8-Bromo-2-naphthol

  • MDL number MFCD06656640
  • Cas Number : 7385-87-7
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA100109
Availability details for
Quantity
  • Availability
  • Add to Cart