ATC311766871 Atlantic Research Chemicals Ltd

4-Chloro-6-iodoquinazoline

  • MDL number MFCD01862193
  • Cas Number : 74173-76-5
  • Molecular Weight : 290.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA100124
Availability details for
Quantity
  • Availability
  • Add to Cart