ATC311766912 Atlantic Research Chemicals Ltd

6-Bromo[1,2,5]thiadiazolo[3,4-b]pyridine

  • MDL number MFCD08446790
  • Cas Number : 72023-75-7
  • Molecular Weight : 216.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA10040
Availability details for
Quantity
  • Availability
  • Add to Cart