ATC311766923 Atlantic Research Chemicals Ltd

5-Bromoimidazo[1,2-a]pyrazine

  • MDL number MFCD08460096
  • Cas Number : 87597-26-0
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA10051
Availability details for
Quantity
  • Availability
  • Add to Cart