ATC311766962 Atlantic Research Chemicals Ltd

6-Bromo[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

  • MDL number MFCD11110678
  • Cas Number : 425702-91-6
  • Molecular Weight : 214.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA10092
Availability details for
Quantity
  • Availability
  • Add to Cart