ATC311766963 Atlantic Research Chemicals Ltd

8-Bromo-1,6-naphthyridine

  • MDL number MFCD00234384
  • Cas Number : 17965-74-1
  • Molecular Weight : 209.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA10093
Availability details for
Quantity
  • Availability
  • Add to Cart