ATC311766981 Atlantic Research Chemicals Ltd

2-Chloro-2,4,4-trimethylpentane

  • MDL number MFCD00101612
  • Cas Number : 6111-88-2
  • Molecular Weight : 148.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA10112
Availability details for
Quantity
  • Availability
  • Add to Cart