ATC311767001 Atlantic Research Chemicals Ltd

(2-Methyl-4-methoxyphenyl)methanol

  • MDL number MFCD04038833
  • Cas Number : 52289-55-1
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HM00119
Availability details for
Quantity
  • Availability
  • Add to Cart