ATC311767012 Atlantic Research Chemicals Ltd

2-Chloro-4-fluorobenzyl alcohol

  • MDL number MFCD00236024
  • Cas Number : 208186-84-9
  • Molecular Weight : 160.574

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=HM00130
Availability details for
Quantity
  • Availability
  • Add to Cart