ATC311767032 Atlantic Research Chemicals Ltd

[2-(Methylamino)pyridin-3-yl]methanol

  • MDL number MFCD06254696
  • Cas Number : 32399-12-5
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HM00217
Availability details for
Quantity
  • Availability
  • Add to Cart