ATC311767109 Atlantic Research Chemicals Ltd

4-(Hydroxymethyl)-1-methylpyrrolidin-2-one

  • MDL number MFCD09055222
  • Cas Number : 59887-20-6
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HM04501
Availability details for
Quantity
  • Availability
  • Add to Cart