ATC311767118 Atlantic Research Chemicals Ltd

4-(Hydroxymethyl)cyclohexanone

  • MDL number MFCD08056334
  • Cas Number : 38580-68-6
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HM09901
Availability details for
Quantity
  • Availability
  • Add to Cart