ATC311767196 Atlantic Research Chemicals Ltd

1-(Bromomethyl)-3-phenoxybenzene

  • MDL number MFCD01761658
  • Cas Number : 51632-16-7
  • Molecular Weight : 263.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=MH00127
Availability details for
Quantity
  • Availability
  • Add to Cart