ATC311767199 Atlantic Research Chemicals Ltd

1-(Bromomethyl)-4-ethoxybenzene

  • MDL number MFCD08444317
  • Cas Number : 2606-57-7
  • Molecular Weight : 215.089

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=MH00130
Availability details for
Quantity
  • Availability
  • Add to Cart