ATC311767208 Atlantic Research Chemicals Ltd

3-Bromo-5-(chloromethyl)pyridine hydrochloride

  • MDL number MFCD09260933
  • Cas Number : 39741-46-3
  • Molecular Weight : 242.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=MH00208
Availability details for
Quantity
  • Availability
  • Add to Cart