ATC311767262 Atlantic Research Chemicals Ltd

5-Chloro-2-(chloromethyl)-1H-benzimidazole

  • MDL number MFCD00060503
  • Cas Number : 20443-38-3
  • Molecular Weight : 201.056

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=MH02202
Availability details for
Quantity
  • Availability
  • Add to Cart