ATC311767268 Atlantic Research Chemicals Ltd

5-(2-Bromoethyl)-2,3-dihydro-1-benzofuran

  • MDL number MFCD06797641
  • Cas Number : 127264-14-6
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=MH03201
Availability details for
Quantity
  • Availability
  • Add to Cart