ATC311767271 Atlantic Research Chemicals Ltd

tert-Butyl 4-(Iodomethyl)piperidine-1-carboxylate

  • MDL number MFCD08437653
  • Cas Number : 145508-94-7
  • Molecular Weight : 325.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=MH03703
Availability details for
Quantity
  • Availability
  • Add to Cart