ATC311767284 Atlantic Research Chemicals Ltd

1-(2-Chloroethyl)azepane hydrochloride

  • MDL number MFCD00012842
  • Cas Number : 26487-67-2
  • Molecular Weight : 198.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=MH09910
Availability details for
Quantity
  • Availability
  • Add to Cart