ATC311767295 Atlantic Research Chemicals Ltd

N-(3-Chloro-2-methylpropyl)-N,N-dimethylamine hydrochloride

  • MDL number MFCD00012514
  • Cas Number : 4261-67-0
  • Molecular Weight : 172.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=MH10102
Availability details for
Quantity
  • Availability
  • Add to Cart