ATC311767362 Atlantic Research Chemicals Ltd

Methyl 5-Chloro-3-chlorosuphonylthiophene-2-carboxylate

  • MDL number MFCD09032981
  • Cas Number : 158439-31-7
  • Molecular Weight : 275.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=SC00314
Availability details for
Quantity
  • Availability
  • Add to Cart