ATC311767435 Atlantic Research Chemicals Ltd

(4-Chloro-3-nitrophenyl)(4-fluorophenyl)methanone

  • MDL number MFCD03840494
  • Cas Number : 31431-16-0
  • Molecular Weight : 279.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001127
Availability details for
Quantity
  • Availability
  • Add to Cart