ATC311767610 Atlantic Research Chemicals Ltd

Cinnamaldehyde thiosemicarbazone

  • MDL number MFCD00170595
  • Cas Number : 5351-70-2
  • Molecular Weight : 205.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001286
Availability details for
Quantity
  • Availability
  • Add to Cart