ATC311767642 Atlantic Research Chemicals Ltd

2-Fluoro-6-(trifluoromethyl)phenol

  • MDL number MFCD00236300
  • Cas Number : 239135-50-3
  • Molecular Weight : 180.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS001315
Availability details for
Quantity
  • Availability
  • Add to Cart