ATC311767671 Atlantic Research Chemicals Ltd

2,6-Difluoro-3-nitrobenzonitrile

  • MDL number MFCD00042254
  • Cas Number : 143879-77-0
  • Molecular Weight : 184.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001341
Availability details for
Quantity
  • Availability
  • Add to Cart