ATC311767693 Atlantic Research Chemicals Ltd

N-[2-(Acetylamino)phenyl]acetamide

  • MDL number MFCD00092752
  • Cas Number : 2050-85-3
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001361
Availability details for
Quantity
  • Availability
  • Add to Cart