ATC311767716 Atlantic Research Chemicals Ltd

(4-Amino-3-bromophenyl)acetonitrile

  • MDL number MFCD08275180
  • Cas Number : 882855-96-1
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001383
Availability details for
Quantity
  • Availability
  • Add to Cart