ATC311767746 Atlantic Research Chemicals Ltd

2-(2-Chlorophenoxy)-N'-hydroxyethanimidamide

  • MDL number MFCD00276991
  • Cas Number : 175276-80-9
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001411
Availability details for
Quantity
  • Availability
  • Add to Cart