ATC311767866 Atlantic Research Chemicals Ltd

5-(4-Nitrophenyl)cyclohexane-1,3-dione

  • MDL number MFCD12963761
  • Cas Number : 55579-75-4
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001520
Availability details for
Quantity
  • Availability
  • Add to Cart