ATC311767870 Atlantic Research Chemicals Ltd

{[(Chloromethyl)thio]methyl}benzene

  • MDL number MFCD02093436
  • Cas Number : 3970-13-6
  • Molecular Weight : 172.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001524
Availability details for
Quantity
  • Availability
  • Add to Cart