ATC311767890 Atlantic Research Chemicals Ltd

4-(4-Oxocyclohexyl)benzonitrile

  • MDL number MFCD06410874
  • Cas Number : 73204-07-6
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001542
Availability details for
Quantity
  • Availability
  • Add to Cart