ATC311767957 Atlantic Research Chemicals Ltd

4-Hydroxybenzenesulphonamide

  • MDL number MFCD00059190
  • Cas Number : 1576-43-8
  • Molecular Weight : 173.191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS001604
Availability details for
Quantity
  • Availability
  • Add to Cart