ATC311767961 Atlantic Research Chemicals Ltd

3,5-Dibromosulphanilamide

  • MDL number MFCD00014783
  • Cas Number : 39150-45-3
  • Molecular Weight : 330

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001608
Availability details for
Quantity
  • Availability
  • Add to Cart