ATC311767964 Atlantic Research Chemicals Ltd

1,2-Dichloro-4-methyl-3-nitro-5-(trifluoromethyl)benzene

  • MDL number MFCD04972788
  • Cas Number : 115571-66-9
  • Molecular Weight : 274.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001610
Availability details for
Quantity
  • Availability
  • Add to Cart