ATC311767992 Atlantic Research Chemicals Ltd

4-(5-Fluoro-2-nitrophenyl)morpholine

  • MDL number MFCD09834357
  • Cas Number : 536977-34-1
  • Molecular Weight : 226.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001636
Availability details for
Quantity
  • Availability
  • Add to Cart