ATC311768023 Atlantic Research Chemicals Ltd

4-Amino-3-methylbenzenesulfonamide

  • MDL number MFCD00466840
  • Cas Number : 53297-70-4
  • Molecular Weight : 186.234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS00176
Availability details for
Quantity
  • Availability
  • Add to Cart