ATC311768057 Atlantic Research Chemicals Ltd

4-Chloro-2,3-dimethylpyridine 1-oxide

  • MDL number MFCD06411125
  • Cas Number : 59886-90-7
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002103
Availability details for
Quantity
  • Availability
  • Add to Cart