ATC311768121 Atlantic Research Chemicals Ltd

6-Piperidin-1-ylnicotinonitrile

  • MDL number MFCD09950318
  • Cas Number : 501378-38-7
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002163
Availability details for
Quantity
  • Availability
  • Add to Cart