ATC311768170 Atlantic Research Chemicals Ltd

2,3,5,6-Tetrachloro-4-(methylsulfonyl)pyridine

  • MDL number MFCD00054553
  • Cas Number : 13108-52-6
  • Molecular Weight : 294.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002208
Availability details for
Quantity
  • Availability
  • Add to Cart